Mouse Ovalbumin Specific IgG1 ELISA Kit
Catalog # ANTIA303480-96
Supplier: ANTIBODIES.COM
New Product
CAS Number:
Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Ovalbumin Specific IgG1
- Description:Mouse ovalbumin specific IgG1 ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:1,563 - 100 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,938 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Ovalbumin Specific IgG1 ELISA kit is a capture Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Ovalbumin Specific IgG1 in serum, plasma, tissue homogenates, and other biological fluids.