Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:BCAT2
- Description:Mouse BCAT2 ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Mouse BCAT2 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse BCAT2. No significant cross-reactivity or interference between BCAT2 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Sensitivity:9,375 pg/ml
- Regulatory status:RUO
Specifications
About this item
Mouse BCAT2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse BCAT2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse BCAT2 ELISA kit (A303475) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse BCAT2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for BCAT2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the BCAT2 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-BCAT2 antibody, which binds the captured BCAT2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of BCAT2 captured in each well. The concentration of BCAT2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.