
Titanium aluminum carbonitride MAX phase, Sigma-Aldrich®
SIAL925950-25G
: Merck
New Product
:
- Pk:25 g
- Seq:3308
- Appearance (Colour):Grey to Black
- Appearance (Form):Powder
- X-Ray Diffraction:Conforms to Structure
- ICP Major Analysis
Confirms Ti and Al Components:Confirmed - Size:Report Result
- Recommended Retest Period:12 Months
:
:M-Clarity™ quality level = MQ100
:GHS09
:196.6 g/mol
:Ti₃AlCN