Titanium aluminium carbide 312 MAX phase ≥90% ≤40 μm, Sigma-Aldrich®
SIAL910775-25G
: Merck
New Product
:
- Pk:25 g
- Seq:3308
- Appearance (Colour):Grey to Black
- Appearance (Form):Powder
- X-Ray Diffraction:Conforms to Structure
- Particle Size (µm):≤40
- Purity:≤90%
:
:194.60 g/mol
:196506-01-1
:Ti₃AlC₂