
Molybdenum titanium aluminum carbide MAX phase, Sigma-Aldrich®
Catalog # SIAL925977-25G
Supplier: Merck
New Product
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance (Colour):Grey to Black
- Appearance (Form):Powder
- X-Ray Diffraction:Conforms to Structure
- ICP Major Analysis
Confirms Mo, Ti and Al Components:Confirmed - Size:Report Result
- Recommended Retest Period:12 Months
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100
:GHS09
:350.63 g/mol
Formula:Mo₂Ti₂AlC₃