ContactUsLinkComponent
Human Untagged LRRC15 Clone
Human Untagged LRRC15 Clone
Catalog # ORIGSC305914
Supplier:  OriGene
New Product
CAS Number:  
Human Untagged LRRC15 Clone
Catalog # ORIGSC305914
Supplier:  OriGene
New Product
CAS Number:  
Specifications
  • Pk:
    10 µG
  • Description:
    Human LRRC15 untagged clone, tag free
  • Vector details:
    pCMV6-Entry
Specifications
About this item
LRRC15 (untagged)-Human leucine rich repeat containing 15 (LRRC15), transcript variant 2.
E. coli selection: Kanamycin (25 µg/ml)
Mammalian cell selection: Neomycin
Insert size: 1700 bp