Order Entry
Czech Republic
ContactUsLinkComponent
Antibiotics Assay Testing Paper Discs
Antibiotics Assay Testing Paper Discs
Catalog # FIOR0905A00005
Supplier:  Ahlstrom Munksjö
CAS Number:  
Antibiotics Assay Testing Paper Discs
Catalog # FIOR0905A00005
Supplier:  Ahlstrom Munksjö
CAS Number:  

Specifications

  • Pk:
    1.000
  • Description:
    Antibiotics testing paper discs 12,7 mm made of 100% cotton linters

Specifications

About this item

Antibiotics testing paper discs 12,7 mm made of 100% cotton linters box of 1000 disks.

  • Weight: 330 g/m²
  • Thickness: 0,9 mm
  • Water absorption: 215 ml
  • Grade 905

Applications: Impregnation of antibiotics for testing the resistance of pathogenic organisms.