Order Entry
Czech Republic
ContactUsLinkComponent
Nexus™ Horizontal Clean Bench, 6' Wide
Nexus™ Horizontal Clean Bench, 6' Wide
Catalog # LBCP5360630
Supplier:  LABCONCO
New Product
Nexus™ Horizontal Clean Bench, 6' Wide
Catalog # LBCP5360630
Supplier:  LABCONCO
New Product

Specifications

  • Description:
    Nexus™ Horizontal clean bench, 1828,8 mm wide, 533,4 mm deep work area with stainless steel standard interior
  • Electrical:
    230 V, 8 A
  • Dimensions:
    191,6×84,4×127,3 mm
  • Weight:
    309,0 kg (shipping)
  • Flow:
    Horizontal
  • Pk:
    1

Specifications

About this item

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications, and meet the rigorous demands of life sciences, materials science applications, pharmaceutical R&D and compounding pharmacies. The Nexus also provides an ISO Class 4 clean airflow environment for protection of critical, non-hazardous samples.

  • Electronically Commutated Motor (ECM)
  • Unidirectional, laminar airflow
  • Constant Airflow Profile™ (CAP) airflow monitoring system
  • Glass side panels
  • Spill-Stopper design protects HEPA filter from spills
  • USB port for data record export
  • Remote connectivity via Ethernet port
  • Prefilters to help extend HEPA filter life
  • Bright, glare-free LED lighting

Laminar airflow is driven by an ECM blower powered with Constant Airflow Profile (CAP™️) technology for continuous, precise airflow that is energy efficient with long-lasting HEPA filters.The sleek Nexus Horizontal Clean Bench design includes tempered safety glass and a stainless-steel interior option for effortless cleaning and durability in critical environments.

The touchscreen interface features Nexus OS™ software that simplifies operation, provides access to user settings and displays detailed alert messages when needed. Operational logs and maintenance records are stored onboard and can be accessed remotely for worry-free compliance management.

Delivery: Base stand included.