Gloves, Nitrile, Powder-Free, Coverfeel Touch
: Medicom
New Product
Kolmi Coverfeel Touch glove Class I - Personal Protective Equipment, nitrile, powder-free and textured fingertips.
- No accelerator to limit type IV allergies
- Ultra-comfortable glove and faster donning thanks to its internal polymer coating
- Great elasticity and good dexterity
- Non-slip texture for excellent grip and handling of objects
- Made in France
Recommended in medical, industrial and agri-food environments.
: EN ISO 374-1 Type B, ASTM D6978-05, EN ISO 374-5:2016.
- Catalog No:
- HOPE1140-B
- HOPE1140-D
- HOPE1140-A
- HOPE1140-C
- HOPE1140-E
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
Frequently Bought Together