Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine 7.5 monoacid
Cyanine 7.5 monoacid [equivalent to Cy7.5® acid]
Cyanine 7.5 monoacid [equivalent to Cy7.5® acid]
Cyanine 7.5 monoacid [equivalent to Cy7.5® acid]
Catalog # AATB167
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
Cyanine 7.5 monoacid [equivalent to Cy7.5® acid]
Catalog # AATB167
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Bal.:
5 mg
Sekvence:
2210
Specifications
About this item
Product Details
:
Storage Temperature:
Freezer
:
1057.10 g/mol
Product Details