Order Entry
Czech Republic
ContactUsLinkComponent
Cyanine 7.5 monosuccinimidyl ester [equivalent to Cy7.5® NHS ester]
Cyanine 7.5 monosuccinimidyl ester [equivalent to Cy7.5® NHS ester]
Catalog # AATB168
Supplier:  AAT Bioquest
New Product
CAS Number:  
Cyanine 7.5 monosuccinimidyl ester [equivalent to Cy7.5® NHS ester]
Catalog # AATB168
Supplier:  AAT Bioquest
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
Storage Temperature:Freezer
 :1343.73 g/mol