Order Entry
Czech Republic
ContactUsLinkComponent
Formaldehyde 1000 µg/ml in water
Health
Formaldehyde 1000 µg/ml in water
Catalog # EHERA13909000WA
Supplier:  EHRENSTORFER
CAS Number:  
Health
Formaldehyde 1000 µg/ml in water
Catalog # EHERA13909000WA
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :30.03 g/mol
 :GHS08
Storage Temperature:Refrigerator
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274