
Hexaamminecobalt trichloride 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10534-89-1
:
Formula:H₁₈CoCl₃N₆
MDL Number:MFCD00036304
:267.47 g/mol
Merck Index:12,04711
Melting Pt:217 °C
Density:1,702 g/cm³ (20 °C)
EINECS:234-103-9
:GHS08
Specification Test Results
- Appearance:Orange to orange-brown Powder
- Total trace metal impurities:≤0.001 %
- Cobalt (Co):21.0 to 23.0 %
Specification Test Results
Frequently Bought Together