Methyl-3,5-dinitrobenzoate, Sigma-Aldrich®
Catalog # SIAL386367-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:226.15 g/mol
:M-Clarity™ quality level = MQ100
EINECS:220-289-9
CAS Number:2702-58-1
Formula:C₈H₆N₂O₆
MDL Number:MFCD00017016