

1,10-Phenanthroline hydrochloride monohydrate 99.5-102% (dried basis) Reag. Ph. Eur. for spectrophotometry, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:18851-33-7
:3829-86-5
Formula:C₁₂H₉ClN₂·1H₂O
MDL Number:MFCD00150061
:234.68 g/mol
Melting Pt:224…227 °C (dec.)
Density:1,385 g/cm³ (-100.15 °C)
Storage Temperature:Ambient
EINECS:223-325-1
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Identity (IR):complying
- Assay:99.5 - 102.0 %
- Melting point:Min. 224 °C
- Loss on drying (105°C):Max. 8 %
- Sulfated ash:Max. 0.2 %
- Sensitive according Fe(II) ions:complying
- Solubility (10 % in H₂O):complying
- Turbidity of solution:complying
- Colour of the solution:complying
Specification Test Results
Frequently Bought Together