Order Entry
Czech Republic
ContactUsLinkComponent
 
Chloroform-[D₁] (99.8% D) for NMR spectroscopy
 
undefined
Chloroform-[D₁] (99.8% D) for NMR spectroscopy
 
 
 

 

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Isotopic enrichment (FT NMR 400 MHz)(D):
    Min. 99.80 %
  • Water:
    Max. 0.01 %

 

Frequently Bought Together