Order Entry
Czech Republic
ContactUsLinkComponent
2,5-Di-n-butoxy-4-morpholinobenzenediazonium p-toluenesulphonate
2,5-Di-n-butoxy-4-morpholinobenzenediazonium p-toluenesulphonate
Catalog # APOSOR98028
Supplier:  Apollo Scientific
CAS Number:  
2,5-Di-n-butoxy-4-morpholinobenzenediazonium p-toluenesulphonate
Catalog # APOSOR98028
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :505.60 g/mol
CAS Number:93672-52-7
Formula:C₂₅H₃₅N₃O₆S