ESD Safety Sandals, Joran Low S1PL
Supplier: ELTEN
New Product
Certificates
About this item
ESD safety shoe made from microfibre/textile upper and a rubber/TPU/EVA sole (Transformers).
- Fabric lining for natural ventilation
- Padded tongue
- Full-length inlay sole ESD
- Non-metallic, penetration-resistant midsole
- Rubber scuff cap
- Composite toe cap
Areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black/red
Certifications: EN ISO 20345 S1PL FO/HRO/SC/SR, form A.
Specifications
- Catalog No:
- ELTN710801/45
- ELTN710801/46
- ELTN710801/47
- ELTN710801/37
- ELTN710801/48
- ELTN710801/38
- ELTN710801/39
- ELTN710801/36
- ELTN710801/40
- ELTN710801/41
- ELTN710801/42
- ELTN710801/43
- ELTN710801/44
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001