Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Violet 540)
Supplier: AAT BIOQUEST
Certificates
About this item
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- AATB10380120
- AATB10380121
- Antigen symbol:
- CD38
- CD38
- Antigen name:
- CD38 antigen
- CD38 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet 540
- mFluor™ Violet 540
- Clone:
- HIT2
- HIT2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together