Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 647)
Supplier: AAT Bioquest
Certificates
About this item
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 647
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB104410F1
- AATB104410F0
- Antigen symbol:
- CD44
- CD44
- Antigen name:
- CD44 antigen
- CD44 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 647
- iFluor® 647
- Clone:
- IM7
- IM7
- Reactivity:
- Human
- Human
- Host:
- Rat
- Rat
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together