Anti-GIGYF1 Rabbit Polyclonal Antibody
Catalog # PRSI55-732
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:PERQ amino acid-rich with GYF domain-containing protein 1
- Antigen symbol:GIGYF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino Acid Range = 666-694
- Form:Liquid
- Gene ID:O75420
- Antigen synonyms:GIGYF1
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:115 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GIGYF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 666-694 amino acids from the Central region of human GIGYF1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
May act cooperatively with GRB10 to regulate tyrosine kinase receptor signaling. May increase IGF1 receptor phosphorylation under IGF1 stimulation as well as phosphorylation of IRS1 and SHC1 (By similarity).
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GIGYF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human