- Antibody type:Primary
- Antigen name:Thy-1 cell surface antigen
- Antigen symbol:THY1
- Clonality:Monoclonal
- Clone:F15-42-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Neutralisation:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P04216
- Antigen synonyms:THY1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Purified human brain CD90 / Thy1 was used as the immunogen for this antibody.
- Tested applications:Functional
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
CD90 (Thy1) is an 18-35 kDa GPI-anchored plasma membrane glycoprotein expressed in many cell types, such as in hematopoietic cells and neurons, connective tissues, various fibroblast and stromal cell lines, tumor endothelial cell lines and others. It is involved in T-cell activation, cellular adhesion, proliferation and migration, neurite outgrowth, wound healing, apoptosis, and fibrosis. CD90 (Thy1) participates in multiple signaling cascades and its effects are tissue- and cell type-specific. It often functions as an important regulator of cell/cell and cell/matrix interactions.
Functional Testing: order BSA/azide-free antibody format Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD90 antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Monkey