Anti-METTL17 Rabbit Polyclonal Antibody
Catalog # PRSI64-217
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Methyltransferase-like protein 17, mitochondrial
- Antigen symbol:METTL17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:Amino Acid Range = 88-116
- Form:Liquid
- Gene ID:64745
- Modification:None
- Molecular weight:51 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This M11D1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 88-116 amino acids from the N-terminal region of human M11D1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 μl
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: METTL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 88-116
Host: Rabbit
Isotype: Ig
Reactivity: Human