Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ULBP2 Rabbit Polyclonal Antibody
Anti-ULBP2 Rabbit Polyclonal Antibody
Catalog # PRSI15-921
Supplier:  ProSci Inc.
Anti-ULBP2 Rabbit Polyclonal Antibody
Catalog # PRSI15-921
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NKG2D ligand 2
  • Antigen symbol:
    ULBP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    80328
  • Antigen synonyms:
    UL16-binding protein 2|NKG2DL2|ULBP2|N2DL2|Retinoic acid early transcript 1H|N2DL-2|ALCAN-alpha|RAET1H|NKG2D ligand 2
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 26-216 of human ULBP2 (NP_079493.1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: ULBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat