Anti-SLC7A1 Rabbit Polyclonal Antibody
Catalog # PRSI15-728
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 7 (cationic amino acid transporter, y+ system), member 1
- Antigen symbol:SLC7A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6541
- Antigen synonyms:ATRC1|REC1L|HCAT1|SLC7A1|CAT-1|ERR
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human SLC7A1
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC7A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat