Anti-MSMB Rabbit Polyclonal Antibody
Catalog # PRSI15-107
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Microseminoprotein beta
- Antigen symbol:MSMB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:4477
- Antigen synonyms:IGBF|PSP|PSP94|PRPS|prostate secreted seminal plasma protein|PSP-94|MSP|beta-microseminoprotein|immunoglobulin binding factor|MSPB|PN44|PSP57|HPC13|seminal plasma beta-inhibin|prostate secretory protein of 94 amino acids
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21-114 of human MSMB (NP_002434.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MSMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human