Anti-SCN2A Rabbit Polyclonal Antibody
Catalog # PRSI13-732
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:sodium voltage-gated channel alpha subunit 2
- Antigen symbol:SCN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6326
- Antigen synonyms:HBA|brain type 2 alpha subunit|sodium channel protein brain II subunit alpha|sodium channel protein type 2 subunit alpha|HBSC II|HBSCII|HBSCI|EIEE11|alpha subunit|Na(v)1.2|voltage-gated sodium channel subunit alpha Nav1.2|type II|BFNIS|alpha 2 polypeptide|NAC2|alpha 1 polypeptide|voltage-gated sodium channel type II alpha subunit|voltage-gated|SCN2A2|SCN2A|BFIS3|sodium channel|sodium channel protein type II subunit alpha|Nav1.21|sodium channel protein|voltage-gated sodium channel subtype II|BFIC3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 275-401 of human SCN2A (NP_001035232.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SCN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat