Anti-SPAG9 Rabbit Polyclonal Antibody
Catalog # PRSI62-796
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:C-Jun-amino-terminal kinase-interacting protein 4
- Antigen symbol:SPAG9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:603-632 aa
- Form:Liquid
- Gene ID:9043
- Modification:None
- Molecular weight:146 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This JLP (SPAG9) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 603-632 amino acids from the central region of human JLP (SPAG9).
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: SPAG9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 603-632
Host: Rabbit
Isotype: Ig
Reactivity: Human