Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # PRSI22-874
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 2 catalytic subunit
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:5563
- Antigen synonyms:catalytic alpha-2 chain|AMPK|5'-AMP-activated protein kinase|AMPK-alpha-2 chain|AMPK2|PRKAA2|protein kinase|PRKAA|alpha 2 catalytic subunit|AMP-activated protein kinase alpha 2 catalytic subunit|AMP-activated
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 343-552 of human PRKAA2 (NP_006243.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat