Anti-ADSS Rabbit Polyclonal Antibody
Catalog # PRSI22-278
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Adenylosuccinate synthetase isozyme 2 {ECO:0000255|HAMAP-Rule:MF_03127}
- Antigen symbol:ADSS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:159
- Antigen synonyms:AdSS 2 {ECO:0000255|HAMAP-Rule:MF_03127}|ADSS {ECO:0000255|HAMAP-Rule:MF_03127}|6344 {ECO:0000255|HAMAP-Rule:MF_03127}|liver isozyme {ECO:0000255|HAMAP-Rule:MF_03127}|Adenylosuccinate synthetase|IMP--aspartate ligase 2 {ECO:0000255|HAMAP-Rule:MF_03127}|L-type adenylosuccinate synthetase {ECO:0000255|HAMAP-Rule:MF_03127}|Adenylosuccinate synthetase isozyme 2 {ECO:0000255|HAMAP-Rule:MF_03127}|AMPSase 2 {ECO:0000255|HAMAP-Rule:MF_03127}|acidic isozyme {ECO:0000255|HAMAP-Rule:MF_03127}
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADSS (NP_001117.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ADSS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat