Anti-HPD Rabbit Polyclonal Antibody
Katalogové číslo PRSI22-273
Dodavatel: ProSci Inc.
New Product
Specifikace
- Antibody type:Primary
- Antigen name:4-hydroxyphenylpyruvate dioxygenase
- Antigen symbol:HPD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:3242
- Antigen synonyms:HPPDase|4HPPD|4-hydroxyphenylpyruvate dioxygenase|HPD|4-hydroxyphenylpyruvic acid oxidase|PPD
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-393 of human HPD (NP_002141.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifikace
O tomto produktu
Type: Primary
Antigen: HPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat