Anti-CES1 Rabbit Polyclonal Antibody
Katalogové číslo PRSI18-347
Dodavatel: ProSci Inc.
New Product
Specifikace
- Antibody type:Primary
- Antigen name:Carboxylesterase 1
- Antigen symbol:CES1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1066
- Antigen synonyms:CES2|CEH|HMSE|SES1|CES1|REH|hCE-1|CE-1|MGC117365|ACAT|HMSE1|TGH|PCE-1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 287-566 of human CES1 (NP_001257,4)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifikace
O tomto produktu
Type: Primary
Antigen: CES1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse