Anti-YTHDF1 Rabbit Polyclonal Antibody
PRSI18-301
- Antibody type:Primary
- Antigen name:YTH N6-methyladenosine RNA binding protein 1
- Antigen symbol:YTHDF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:54915
- Antigen synonyms:C20orf21|YTH domain family protein 1|YTH domain family|YTH domain-containing family protein 1|DACA-1|member 1|YTH N(6)-methyladenosine RNA binding protein 1|dermatomyositis associated with cancer putative autoantigen 1|YTH domain family 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of human YTHDF1
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: YTHDF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat