Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-SLC6A9 Rabbit Polyclonal Antibody
Catalog # PRSI16-490
Supplier:  ProSci Inc.
Anti-SLC6A9 Rabbit Polyclonal Antibody
Catalog # PRSI16-490
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute carrier family 6 member 9
  • Antigen symbol:
    SLC6A9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    6536
  • Antigen synonyms:
    GCENSG|GLYT1|glyT-1|glycine)|sodium- and chloride-dependent glycine transporter 1|solute carrier family 6 (neurotransmitter transporter|member 9
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human SLC6A9 (NP_964012,2)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: SLC6A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat