Anti-SLC14A1 Rabbit Polyclonal Antibody
PRSI16-368
- Antibody type:Primary
- Antigen name:Solute carrier family 14 (urea transporter), member 1 (Kidd blood group)
- Antigen symbol:SLC14A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6563
- Antigen synonyms:FLJ33745|UTE|SLC14A1|UT1|RACH2|JK|HUT11|HsT1341|UT-B1|RACH1|FLJ41687
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human SLC14A1 (NP_001295208,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC14A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat