Anti-POLR3C Rabbit Polyclonal Antibody
PRSI16-230
- Antibody type:Primary
- Antigen name:DNA-directed RNA polymerase III subunit RPC3
- Antigen symbol:POLR3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10623
- Antigen synonyms:DNA-directed RNA polymerase III subunit RPC3|DNA-directed RNA polymerase III subunit C|RPC62|POLR3C|RNA polymerase III 62 kDa subunit|RNA polymerase III subunit C3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human POLR3C (NP_006459,3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: POLR3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat