Anti-SLC1A3 Rabbit Polyclonal Antibody
PRSI16-199
- Antibody type:Primary
- Antigen name:Solute carrier family 1 (glial high affinity glutamate transporter), member 3
- Antigen symbol:SLC1A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6507
- Antigen synonyms:OTTHUMP00000116002|solute carrier family 1 (glial high affinity glutamate transporter)|SLC1A3|EA6|GLAST|solute carrie|GLAST1|GLAST-1|sodium-dependent glutamate/aspartate transporter 1|FLJ25094|member 3|EAAT1|excitatory amino acid transporter 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 453-542 of human SLC1A3 (NP_004163,3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat