Anti-CISH Rabbit Polyclonal Antibody
Katalogové číslo PRSI22-101
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cytokine inducible SH2 containing protein
- Antigen symbol:CISH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1154
- Antigen synonyms:BACTS2|G18|suppressor of cytokine signaling|cytokine-inducible inhibitor of signaling type 1B|SOCS|CIS-1|cytokine-inducible SH2-containing protein|CIS
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-257 of human CISH (NP_659508.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CISH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat