Anti-PIWIL2 Rabbit Polyclonal Antibody
PRSI19-986
New Product
- Antibody type:Primary
- Antigen name:Piwi-like RNA-mediated gene silencing 2
- Antigen symbol:PIWIL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:57746
- Antigen synonyms:Miwi like|piwi-like homolog 2|piwi-like protein 2|mili|Miwi likw|Piwil1l
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of mouse PIWIL2.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PIWIL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat