Anti-CYP2A6 Rabbit Polyclonal Antibody
Catalog # PRSI19-876
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450 family 2 subfamily A member 6
- Antigen symbol:CYP2A6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1548
- Antigen synonyms:subfamily A|cytochrome P450(I)|P450C2A|CPA6|coumarin 7-hydroxylase|CYPIIA6|polypeptide 6|cytochrome P45family 2|P450PB|1,4-cineole 2-exo-monooxygenase|cytochrome P450 2A6|xenobiotic monooxygenase|CYP2A3|cytochrome P450 IIA3|CYP2A|flavoprotein-linked monooxygenase|cytochrome P45subfamily IIA (phenobarbital-inducible)
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 215-494 of human CYP2A6 (NP_000753.3).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CYP2A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse