Anti-HSD17B10 Rabbit Polyclonal Antibody
PRSI19-669
- Antibody type:Primary
- Antigen name:Hydroxysteroid (17-beta) dehydrogenase 10
- Antigen symbol:HSD17B10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:3028
- Antigen synonyms:ERAB|17b-HSD10|AB-binding alcohol dehydrogenase|HADH2|MRPP2|XH98G2|MHBD|HSD17B10|type II|type 10 17b-HSD|type 10 17beta-hydroxysteroid dehydrogenase|SCHAD|ABAD|hydroxyacyl-Coenzyme A dehydrogenase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-261 of human HSD17B10 (NP_004484.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: HSD17B10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat