Anti-SLC1A6 Rabbit Polyclonal Antibody
PRSI18-863
- Antibody type:Primary
- Antigen name:Solute carrier family 1 member 6
- Antigen symbol:SLC1A6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6511
- Antigen synonyms:member 6|sodium-dependent glutamate/aspartate transporter|solute carrier family 1 (high affinity aspartate/glutamate transporter)|EAAT4|excitatory amino acid transporter 4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 155-265 of human SLC1A6 (NP_001259016.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC1A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat