Anti-KLHL4 Rabbit Polyclonal Antibody
Catalog # PRSI60-089
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Kelch-like protein 4
- Antigen symbol:KLHL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:84 - 113 aa
- Form:Liquid
- Gene ID:56062
- Modification:None
- Molecular weight:80 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4°C
- Immunogen:This KLHL4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 84 - 113 amino acids from the N-terminal region of human KLHL4.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: KLHL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 84-113
Host: Rabbit
Isotype: Ig
Reactivity: Human