Anti-AKR1E2 Rabbit Polyclonal Antibody
Katalogové číslo PRSI60-077
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:1,5-anhydro-D-fructose reductase
- Antigen symbol:AKR1E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:291 - 320 aa
- Form:Liquid
- Gene ID:83592
- Modification:None
- Molecular weight:37 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This AKR1E2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 291 - 320 amino acids from the C-terminal region of human AKR1E2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: AKR1E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 291-320
Host: Rabbit
Isotype: Ig
Reactivity: Human