Anti-MS4A14 Rabbit Polyclonal Antibody
Katalogové číslo PRSI59-698
Dodavatel: ProSci Inc.
Specifikace
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-domains subfamily A member 14
- Antigen symbol:MS4A14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:556 - 585 aa
- Form:Liquid
- Gene ID:84689
- Modification:None
- Molecular weight:77 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This MS4A14 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 556 - 585 amino acids from the C-terminal region of human MS4A14.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifikace
O tomto produktu
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MS4A14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 556-585
Host: Rabbit
Isotype: Ig
Reactivity: Human