Anti-OSGIN1 Rabbit Polyclonal Antibody
PRSI59-621
New Product
- Antibody type:Primary
- Antigen name:Oxidative stress-induced growth inhibitor 1
- Antigen symbol:OSGIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:483 - 510 aa
- Form:Liquid
- Gene ID:29948
- Modification:None
- Molecular weight:61 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,43 mg/ml
- Shipping temperature:4°C
- Immunogen:This OSGIN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 483 - 510 amino acids from the C-terminal region of human OSGIN1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: OSGIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 483-510
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse