Anti-MMADHC Rabbit Polyclonal Antibody
PRSI59-618
- Antibody type:Primary
- Antigen name:Methylmalonic aciduria and homocystinuria type D protein, mitochondrial
- Antigen symbol:MMADHC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:226 - 253 aa
- Form:Liquid
- Gene ID:27249
- Modification:None
- Molecular weight:33 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This MMADHC antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 226 - 253 amino acids from the C-terminal region of human MMADHC.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MMADHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 226-253
Host: Rabbit
Isotype: Ig
Reactivity: Human