Anti-ALG5 Rabbit Polyclonal Antibody
PRSI59-493
New Product
- Antibody type:Primary
- Antigen name:Dolichyl-phosphate beta-glucosyltransferase
- Antigen symbol:ALG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:74 - 100 aa
- Form:Liquid
- Gene ID:29880
- Modification:None
- Molecular weight:37 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This ALG5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 74 - 100 amino acids from the Central region of human ALG5.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ALG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 74-100
Host: Rabbit
Isotype: Ig
Reactivity: Human