Anti-HLA-C Rabbit Polyclonal Antibody
PRSI59-490
- Antibody type:Primary
- Antigen name:HLA class I histocompatibility antigen, Cw-7 alpha chain
- Antigen symbol:HLA-C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:66 - 93 aa
- Form:Liquid
- Gene ID:3107
- Modification:None
- Molecular weight:41 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This HLA - C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 66 - 93 amino acids from the Central region of human HLA - C.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: HLA-C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 66-93
Host: Rabbit
Isotype: Ig
Reactivity: Human