Anti-MON1B Rabbit Polyclonal Antibody
Catalog # PRSI59-430
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Vacuolar fusion protein MON1 homolog B
- Antigen symbol:MON1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:396 - 425 aa
- Form:Liquid
- Gene ID:22879
- Modification:None
- Molecular weight:59 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,35 mg/ml
- Shipping temperature:4°C
- Immunogen:This MON1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 396 - 425 amino acids from the C-terminal region of human MON1B.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MON1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 396-425
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse